Learn More
Brookfield Engineer Labs Jerome J505-Mercury Vapor Analyzer
Supplier:  Brookfield Engineer Labs J5050005
Encompass
The Jerome® J505 Mercury Vapor Analyzer is a portable fluorescence spectroscopy analyzer, which allows the detection cell to be simpler, smaller, lighter weight and more durable than competing spectroscopy instruments. The highly efficient optical cell requires less flow to purge the system, allowing the J505 to run at a lower flow rate minimizing sample dilution as found in competing spectroscopy instruments.
Catalog No. NC2960814
Add to Cart
Request a Quote