Learn More
ATCC J558; Plasmacytoma; Mouse (Mus musculus)
Catalog No. 502384698
Encompass
J558 are B lymphocytes that were isolated from a mouse with plasmacytoma. It has applications in immune system disorder research and immunology.
Catalog No. 50-238-4698 Supplier ATCC Supplier No. TIB6
J558 are B lymphocytes that were isolated from a mouse with plasmacytoma. It has applications in immune system disorder research and immunology.