Learn More
Rudolph Research Analytical Rudolph J57(S5) Refractometer, Electronic Peltier Temperature Control, Environmental Protection (EP) Presser
SDP
Catalog No. NC2942685
Encompass
The Rudolph J57(S5) Refractometer with Environmental Protection (EP) Presser and Variable Temperature (VT) is designed for precision in challenging environments. Offering temperature control from 10 to 65 °C, it ensures accurate refractive index and Brix measurements while safeguarding against moisture and sunlight. Ideal for lab applications in food, beverage, and pharmaceuticals, this automatic refractometer combines user-friendly operation with high durability, making it a reliable choice.
Catalog No. NC2942685 Supplier Rudolph Research Analytical Supplier No. J57S5
Add to Cart
Add to Cart
Request a Quote
The Rudolph J57(S5) Refractometer with Environmental Protection (EP) Presser and Variable Temperature (VT) is designed for precision in challenging environments. Offering temperature control from 10 to 65 °C, it ensures accurate refractive index and Brix measurements while safeguarding against moisture and sunlight. Ideal for lab applications in food, beverage, and pharmaceuticals, this automatic refractometer combines user-friendly operation with high durability, making it a reliable choice.