Learn More
Beckman Coulter Beckman Coulter JA-14.50 Fixed-Angle Rotor, 16 x 50 mL, single-locking lid (B88555)
Catalog No. NC1973082
Encompass
The Beckman Coulter JA-14.50 (B88555) fixed-angle rotor is engineered for high-throughput processing of 16 x 50 mL conical tubes. It delivers up to 14,000 rpm (35,100 x g) for pelleting and density gradient separations, and features a single-locking lid and lightweight construction for easier handling.
  • Holds 16 x 50 mL conical tubes.
  • Fixed-angle design for efficient pelleting and density gradient separations.
  • Maximum speed 14,000 rpm; maximum RCF 35,100 x g.
  • Single-locking lid for secure operation.
  • Lightweight construction for easier handling and installation.
  • Designed for use in high-speed refrigerated centrifuges.
Catalog No. NC1973082 Supplier Beckman Coulter Supplier No. B88555
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Beckman Coulter JA-14.50 (B88555) fixed-angle rotor is engineered for high-throughput processing of 16 x 50 mL conical tubes. It delivers up to 14,000 rpm (35,100 x g) for pelleting and density gradient separations, and features a single-locking lid and lightweight construction for easier handling.
  • Holds 16 x 50 mL conical tubes.
  • Fixed-angle design for efficient pelleting and density gradient separations.
  • Maximum speed 14,000 rpm; maximum RCF 35,100 x g.
  • Single-locking lid for secure operation.
  • Lightweight construction for easier handling and installation.
  • Designed for use in high-speed refrigerated centrifuges.