Learn More
Beckman Coulter Beckman Coulter JA-18 Fixed-Angle Rotor, 10 x 100 mL, 18,000 rpm
Catalog No. NC2410616 Shop All Beckman Coulter Products
Encompass
High-capacity fixed-angle rotor for Avanti-series centrifuges, designed for large-volume, high-speed pelleting applications including bacteria, cell membranes, and subcellular organelles. Holds ten 100 mL tubes and operates up to 18,000 rpm (47,900 x g); compatible with Avanti J-26S, J-E, and J-30I centrifuges.
  • Holds 10 x 100 mL tubes.
  • Operates to 18,000 rpm (47,900 x g).
  • K factor 566 for sedimentation efficiency.
  • Suitable for pelleting bacteria, cell membranes, and organelles.
  • Independently certified to contain liquids and aerosols when used and maintained properly.
Catalog No. NC2410616 Supplier Beckman Coulter Supplier No. 369679
Add to Cart
Add to Cart
Request a Quote
High-capacity fixed-angle rotor for Avanti-series centrifuges, designed for large-volume, high-speed pelleting applications including bacteria, cell membranes, and subcellular organelles. Holds ten 100 mL tubes and operates up to 18,000 rpm (47,900 x g); compatible with Avanti J-26S, J-E, and J-30I centrifuges.
  • Holds 10 x 100 mL tubes.
  • Operates to 18,000 rpm (47,900 x g).
  • K factor 566 for sedimentation efficiency.
  • Suitable for pelleting bacteria, cell membranes, and organelles.
  • Independently certified to contain liquids and aerosols when used and maintained properly.