Learn More
Medchemexpress LLC JA-ACC-d3 (Jasmonyl-ACC-d3) | 371778-55-1 | C16H20D3NO4 | 1 MG
SDP
Catalog No. 5000373682
Encompass_Preferred
JA-ACC-d3 is deuterium labeled JA-ACC, primarily for research purposes. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-73682 Supplier Medchemexpress LLC Supplier No. HY151931S1MG
JA-ACC-d3 is deuterium labeled JA-ACC, primarily for research purposes. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles