Learn More
Beckman Coulter Beckman Coulter JA-14.50 Fixed-Angle Aluminum Rotor, 16 x 50 mL conical tubes - for Avanti J Series centrifuges (MPN B88554)
Catalog No. NC2331922
Encompass
The Beckman Coulter JA-14.50 fixed-angle aluminum rotor (MPN B88554) is a BioSafe rotor designed for use with Avanti J Series centrifuges. It supports pelleting, density-gradient separations, and routine high-throughput centrifugation of biological samples.
  • Holds sixteen 50 mL conical tubes.
  • 35° fixed-angle design for compact pellets and efficient separation.
  • Maximum speed 14,000 rpm for accelerated processing.
  • k-factor 787 for performance characterization.
  • Aluminum construction with BioSafe design for routine laboratory use.
  • Nominal capacity 800 mL, rMIN 87 mm, rMAX 160 mm.
Catalog No. NC2331922 Supplier Beckman Coulter Supplier No. B88554
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Beckman Coulter JA-14.50 fixed-angle aluminum rotor (MPN B88554) is a BioSafe rotor designed for use with Avanti J Series centrifuges. It supports pelleting, density-gradient separations, and routine high-throughput centrifugation of biological samples.
  • Holds sixteen 50 mL conical tubes.
  • 35° fixed-angle design for compact pellets and efficient separation.
  • Maximum speed 14,000 rpm for accelerated processing.
  • k-factor 787 for performance characterization.
  • Aluminum construction with BioSafe design for routine laboratory use.
  • Nominal capacity 800 mL, rMIN 87 mm, rMAX 160 mm.