Learn More
Beckman Coulter JA25.50 FIXED ANGLE, 8 X 50ML
Catalog No. NC1587943
Encompass
JA25.50 fixed angle, 8 x 50mL rotor, Dual-Locking lid, 25,000 rpm, 75,600 x g
Catalog No. NC1587943 Supplier Beckman Coulter Supplier No. 363058
Add to Cart
Add to Cart
Request a Quote
JA25.50 fixed angle, 8 x 50mL rotor, Dual-Locking lid, 25,000 rpm, 75,600 x g