Learn More
Beckman Coulter Beckman Coulter JA-25.50 Fixed-Angle Rotor, Aluminum, 8 x 50 mL, 25,000 rpm (75,600 x g)
Catalog No. NC1702810
Encompass
The JA-25.50 fixed-angle rotor is an anodized aluminum rotor designed for high-speed preparative centrifugation, providing reliable capacity for eight 50 mL tubes and compatibility with Avanti J-series centrifuges.
  • Capacity for eight 50 mL tubes.
  • Fixed-angle design for efficient pelleting.
  • Anodized aluminum construction for durability.
  • Maximum speed 25,000 rpm (75,600 x g) for high-force applications.
  • Suitable for harvesting cells, processing tissue homogenates, and subcellular separations.
Catalog No. NC1702810 Supplier Beckman Coulter Supplier No. 363055
Add to Cart
Add to Cart
Request a Quote
The JA-25.50 fixed-angle rotor is an anodized aluminum rotor designed for high-speed preparative centrifugation, providing reliable capacity for eight 50 mL tubes and compatibility with Avanti J-series centrifuges.
  • Capacity for eight 50 mL tubes.
  • Fixed-angle design for efficient pelleting.
  • Anodized aluminum construction for durability.
  • Maximum speed 25,000 rpm (75,600 x g) for high-force applications.
  • Suitable for harvesting cells, processing tissue homogenates, and subcellular separations.