Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JAB-3068 (hydrochloride) | 2169223-49-6 | 98.9% | 100 MG
SDP

Catalog No. 5000338939
Encompass_Preferred

JAB-3068 hydrochloride is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM.

  • Potent SHP2 inhibitor
  • Inhibits proliferation of KYSE-520 cells
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight of 513.00
  • Chemical formula C22H27ClF2N6O2S

Catalog No. 50-003-38939 Supplier Medchemexpress LLC Supplier No. HY131132A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JAB-3068 hydrochloride is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM.

  • Potent SHP2 inhibitor
  • Inhibits proliferation of KYSE-520 cells
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight of 513.00
  • Chemical formula C22H27ClF2N6O2S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.