Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JAB-3068 hydrochloride | 2169223-49-6 | 98.9% | 50 MG
SDP

Catalog No. 5000338555
Encompass_Preferred

JAB-3068 hydrochloride (SHP2-IN-6 hydrochloride) is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM. This compound is a solid with a light yellow to yellow appearance and a molecular weight of 513.00.

  • Potent SHP2 inhibitor
  • Inhibits proliferation of KYSE-520 cells
  • Molecular weight: 513.00
  • Chemical formula: C22H27ClF2N6O2S
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Solubility in DMSO: 115 mg/mL (224.17 mM)

Catalog No. 50-003-38555 Supplier Medchemexpress LLC Supplier No. HY131132A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JAB-3068 hydrochloride (SHP2-IN-6 hydrochloride) is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM. This compound is a solid with a light yellow to yellow appearance and a molecular weight of 513.00.

  • Potent SHP2 inhibitor
  • Inhibits proliferation of KYSE-520 cells
  • Molecular weight: 513.00
  • Chemical formula: C22H27ClF2N6O2S
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Solubility in DMSO: 115 mg/mL (224.17 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.