Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAC Medical JAC-Excel Midsize Phlebotomy Tray Units

Catalog No. 22038446
Change view
Click to view available options
Holds:
42+ tubes for blood draws
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-038446 42+ tubes for blood draws
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-038446 Supplier JAC Medical Supplier No. JAC 400H

missing translation for 'validMainframeMsg'

For medium to small volume blood draws

  • Blue, autoclavable polypropylene with overhead handle
  • 15 × 10.625 × 3 in. (38.1 × 27 × 7.6cm)

Midsize Tray Unit for 42+ Tubes (Cat. No. 22-038-446)

  • Supply organizer
  • Packet holder
  • Glove holder
  • Isolation box w/supply organizer for 42+ Vac tubes
  • Pen holder
  • Clipboard
Midsize Tray Unit for 26+ Tubes (Cat. No. 22-038-448)
  • 26-Well test tube rack
  • Slipholder
  • Two mini utility boxes
  • Two utility boxes
  • Packet holder
  • Glove holder
  • Isolation box
  • Pen holder
  • Clipboard

Specifications

Holds 42+ tubes for blood draws
Autoclavable Autoclavable
Height (English) Handle 8.25 in.
Product Type Phlebotomy Tray Unit
Height (Metric) Handle 20.95cm
Includes Overhead handle, supply organizer, packet holder, glove holder, isolation box, pen holder, clipboard
Material Polypropylene
Size Medium
For Use With (Application) For medium to small volume Blood draws
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.