Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAC Medical Hematology Test Tube Rack

Catalog No. 22038380
Change view
Click to view available options
Includes:
White acrylic base
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-038-380 White acrylic base
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-038-380 Supplier JAC Medical Supplier No. JAC 2

missing translation for 'validMainframeMsg'

For use with JAC Medical phlebotomy trays, isolation box, or countertop

  • Made of clear PETG plastic with a strong white acrylic base

Specifications

No. of Places 20.0
For Use With (Equipment) For phlebotomy tray, isolation box, or countertop, 13mm tubes
Material PETG
Product Type Hematology Test Tube Rack
Color Clear
Includes White acrylic base
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.