Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAC Medical Holders

Catalog No. 22038383
Change view
Click to view available options
For Use With (Equipment):
For phlebotomy tray
Includes:
Two removable divider
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
22-038383 For phlebotomy tray Two removable divider
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-038383 Supplier JAC Medical Supplier No. JAC 5

missing translation for 'validMainframeMsg'

Specifications

Autoclavable Autoclavable
For Use With (Equipment) For phlebotomy tray
For Use With (Application) Provides one, two/three compartments of various size
Color White
Product Type Needle/Vac Tube Holder
Holds 32+ vac tube
Includes Two removable divider
Material Polypropylene
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.