Learn More
Bioworld JACALIN COATED MICROPLATES
Catalog No. 50197549
Encompass
Jacalin, belonging to the lectins family, is a Hemagglutinin from the seeds of the jack fruit Artrocarpus integrifolia. It is well known that lectins have been used extensively for the isolation of glyco-conjugates and glycoproteins with...
Catalog No. 50-197-549 Supplier Bioworld Supplier No. 201400181
Jacalin, belonging to the lectins family, is a Hemagglutinin from the seeds of the jack fruit Artrocarpus integrifolia. It is well known that lectins have been used extensively for the isolation of glyco-conjugates and glycoproteins with...