Learn More
Cayman Chemical JacarIc AcId 10mg
SDP
Catalog No. 502971263
Encompass_Preferred
A conjugated PUFA that induces cell cycle arrest and apoptosis in a variety of cancer cell lines (GI50 = 1-5 µM); increases the production of ROS, leading to oxidative stress that contributes to cytotoxicity
Catalog No. 50-297-1263 Supplier Cayman Chemical Supplier No. 1603610 MG
A conjugated PUFA that induces cell cycle arrest and apoptosis in a variety of cancer cell lines (GI50 = 1-5 µM); increases the production of ROS, leading to oxidative stress that contributes to cytotoxicity