Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jack Bean Meal, Powder, Spectrum™ Chemical
SDP

Catalog No. 18603610 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-610 100 g Poly Bottle
18-603-611 500 g Poly Bottle
18-603-612 2.5 kg Poly Pail
18-603-613 10 kg Poly Pail
18-603-614 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-610 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. JA105100GM
Add to Cart
Add to Cart

JA105, 9002-13-5

Spectrum™ Chemical Jack Bean Meal, Powder is a perennial climbing legume usually cultivated as an annual. Its flowers are pink or white with a red base and pds can be up to 36 cm long. The plant, pods and seeds are used to feed animals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9002-13-5

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 100 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.