Learn More
Chemglass Life Sciences Jacketed Beaker, 200 x 100mm ID, 250 x 120mm OD
SDP
Catalog No. 50872934
Encompass_Preferred
Jacketed Beaker, 200 x 100mm ID, 250 x 120mm OD. Similar to CG-1103, but with a larger opening at the top of the jacket and a shorter overall height. Width to Height ratio on all sizes is 2:1. Beakers also have a FLAT Bottom on both the inner and outer vessel. Hose Connections have an O.D. of 10mm at the largest serration.
Catalog No. 50-872-934 Supplier Chemglass Life Sciences Supplier No. CG110712
Jacketed Beaker, 200 x 100mm ID, 250 x 120mm OD. Similar to CG-1103, but with a larger opening at the top of the jacket and a shorter overall height. Width to Height ratio on all sizes is 2:1. Beakers also have a FLAT Bottom on both the inner and outer vessel. Hose Connections have an O.D. of 10mm at the largest serration.