Learn More
Chemglass Life Sciences Jacketed Beaker, 150 x 75mm ID, 190 x 95mm OD
SDP
Catalog No. 50872933
Encompass_Preferred
Jacketed Beaker, 150 x 75mm ID, 190 x 95mm OD. Similar to CG-1103, but with a larger opening at the top of the jacket and a shorter overall height. Width to Height ratio on all sizes is 2:1. Beakers also have a FLAT Bottom on both the inner and outer vessel. Hose Connections have an O.D. of 10mm at the largest serration.
Catalog No. 50-872-933 Supplier Chemglass Life Sciences Supplier No. CG110711
Jacketed Beaker, 150 x 75mm ID, 190 x 95mm OD. Similar to CG-1103, but with a larger opening at the top of the jacket and a shorter overall height. Width to Height ratio on all sizes is 2:1. Beakers also have a FLAT Bottom on both the inner and outer vessel. Hose Connections have an O.D. of 10mm at the largest serration.