Learn More
Sigma Aldrich Fine Chemicals Biosciences Jacketed beaker1EA
Catalog No. 502905442
Encompass
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.
Catalog No. 50-290-5442 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2027381EA
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.