Learn More
Synthware™ Jacketed Vigreux Distillation Head- Microscale
GREENER_CHOICESDP
Catalog No. 31501053 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
Catalog No. Standard Taper Joint
31-501-053 14/20
1 options
Catalog No. 31-501-053 Supplier Synthware™ Supplier No. D171014
Add to Cart
Add to Cart
Kacketed Vigreux column to improve distillation performance.
Specifications
For Use With (Application) Organic Synthesis
Type Jacketed Vigreux Distillation Head
Material Borosilicate Glass
For Use With (Equipment) D201414 Distribution Adapter
Quantity 1 Ea.
Standard Taper Joint 14/20
Joint Type Ground Glass
Size Microscale