Learn More
MSC PRO-SAFE Jacket, Large, Green, Cotton, Flame-Resistant
Catalog No. 502835161
Encompass
This flame-resistant cotton jacket is designed for protective wear in industrial environments. This large-sized green jacket is made from 9.00 oz cotton and features snap closures, providing comfort and protection against hazards such as slag and grinder sparks.
  • Made from flame-resistant, flame-retardant cotton
  • Provides protection against slag and grinder sparks
  • Lightweight design for comfort
  • Offers good coverage
  • Features snap closures
Catalog No. 50-283-5161 Supplier MSC Supplier No. 42649707
This flame-resistant cotton jacket is designed for protective wear in industrial environments. This large-sized green jacket is made from 9.00 oz cotton and features snap closures, providing comfort and protection against hazards such as slag and grinder sparks.
  • Made from flame-resistant, flame-retardant cotton
  • Provides protection against slag and grinder sparks
  • Lightweight design for comfort
  • Offers good coverage
  • Features snap closures