Learn More
MSC PRO-SAFE Jacket, Medium, Cotton, Flame-Resistant
Catalog No. 502835222
Encompass
The PRO-SAFE flame-resistant cotton jacket is designed for professional use, offering protection and durability. It features a snaps closure and is suitable for environments requiring flame-resistant apparel.
  • Green color
  • Snaps closure
  • Medium size
  • Flame-resistant and flame-retardant
  • Cotton material
  • Chest size 38 to 40 inch
  • Material weight 9.00 oz
Catalog No. 50-283-5222 Supplier MSC Supplier No. 42649293
The PRO-SAFE flame-resistant cotton jacket is designed for professional use, offering protection and durability. It features a snaps closure and is suitable for environments requiring flame-resistant apparel.
  • Green color
  • Snaps closure
  • Medium size
  • Flame-resistant and flame-retardant
  • Cotton material
  • Chest size 38 to 40 inch
  • Material weight 9.00 oz