Learn More
Med Vet International Jackson Cat Catheter, 4Fr x 5.5in, J0153JB, Each
Catalog No. 5000583073
Encompass_Preferred
This specially designed feline catheter is for aid in treatment of FLUTD. It is a modification of the standard feline catheter but with a shorter length (11cm) and metal stylette. There are convenient holes in the luer mount for suture attachment. Sterile.
Catalog No. 50-005-83073 Supplier Med Vet International Supplier No. JOR153JB
This specially designed feline catheter is for aid in treatment of FLUTD. It is a modification of the standard feline catheter but with a shorter length (11cm) and metal stylette. There are convenient holes in the luer mount for suture attachment. Sterile.