Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jackson Immuno Research Labs Jackson ImmunoResearch Secondary Antibody, HRP, F(ab')₂ Fragment, Goat Anti-Human IgG (H+L)
SDP

Catalog No. NC3984990
Encompass_Preferred

This HRP-conjugated F(ab')₂ fragment goat anti-human IgG (H+L) secondary antibody is for in vitro research. It reacts with whole molecule human IgG and light chains of other human immunoglobulins. F(ab')₂ fragments avoid binding to Fc receptors on live cells or to Protein A/G.

  • Target: Human
  • Host: Goat
  • Antibody format: F(ab')₂ Fragment
  • Specificity: IgG (H+L)
  • Conjugate: HRP
  • Clonality: Polyclonal
  • Purity: Purified via pepsin digestion and immunoaffinity chromatography.
  • Usage: Immunohistochemistry, Western blotting, and ELISA.
  • Reduced background for mammalian cell immunostaining.
  • Physical state: Freeze-dried solid.
  • Storage (freeze-dried): 2–8°C.
  • Working concentration: 1:500 to 1:200,000 across applications.

Catalog No. NC3984990 Supplier Jackson Immuno Research Labs Supplier No. 109036003
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This HRP-conjugated F(ab')₂ fragment goat anti-human IgG (H+L) secondary antibody is for in vitro research. It reacts with whole molecule human IgG and light chains of other human immunoglobulins. F(ab')₂ fragments avoid binding to Fc receptors on live cells or to Protein A/G.

  • Target: Human
  • Host: Goat
  • Antibody format: F(ab')₂ Fragment
  • Specificity: IgG (H+L)
  • Conjugate: HRP
  • Clonality: Polyclonal
  • Purity: Purified via pepsin digestion and immunoaffinity chromatography.
  • Usage: Immunohistochemistry, Western blotting, and ELISA.
  • Reduced background for mammalian cell immunostaining.
  • Physical state: Freeze-dried solid.
  • Storage (freeze-dried): 2–8°C.
  • Working concentration: 1:500 to 1:200,000 across applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.