Learn More
MSC Jackson Safety Cap Shield Adapter, 437, Quick Detach, 1 ea
Catalog No. 5000803652
Encompass
This cap shield adapter is designed for use with hard hats, providing a secure method to attach face shields. It features an all-plastic, dielectric construction for enhanced safety and durability.
  • Quick detach brackets
  • Polyethylene spark deflector
  • Totally dielectric, all plastic device for slotted caps
  • Face shields are secured with integral plastic lugs
  • Curved plastic channel fits over the bill of the cap
  • Compatible with SC-6 and Sentry III hard hat styles
Catalog No. 50-008-03652 Supplier MSC Supplier No. 53257085
This cap shield adapter is designed for use with hard hats, providing a secure method to attach face shields. It features an all-plastic, dielectric construction for enhanced safety and durability.
  • Quick detach brackets
  • Polyethylene spark deflector
  • Totally dielectric, all plastic device for slotted caps
  • Face shields are secured with integral plastic lugs
  • Curved plastic channel fits over the bill of the cap
  • Compatible with SC-6 and Sentry III hard hat styles