Learn More
MSC Jackson Safety Face Shield, Clear Acetate, 9 in., 1 ea
Catalog No. 5000686853
Encompass
This acetate face shield offers protection for chemical splash applications. It features excellent flexibility and meets ANSI Z87.1-2010 standards.
  • Excellent flexibility
  • Meets ANSI Z87.1-2010 standards
  • Acetate material
  • Clear lens
  • Nine in. high
  • Has 0.04 in. thickness
Catalog No. 50-006-86853 Supplier MSC Supplier No. 02484095
This acetate face shield offers protection for chemical splash applications. It features excellent flexibility and meets ANSI Z87.1-2010 standards.
  • Excellent flexibility
  • Meets ANSI Z87.1-2010 standards
  • Acetate material
  • Clear lens
  • Nine in. high
  • Has 0.04 in. thickness