Learn More
MSC Jackson Safety Internal Safety Lens, 5-1/4 in, 1 ea
Catalog No. 5000766551
Encompass
The Jackson Safety Internal Safety Lens is a genuine replacement part designed for use with Jackson TrueSight and TrueSight II auto-darkening welding lenses. It protects the auto-darkening filter (ADF) from sparks, scratches, and smoke, extending its lifespan and maintaining clear visibility during welding operations.
  • Works with TrueSight and TrueSight II digital ADF welding helmets
  • Extends the life of your ADF filter
  • Helps protect from sparks, scratches, and smoke
  • Maintains clear visibility for welding tasks
  • Genuine replacement part
Catalog No. 50-007-66551 Supplier MSC Supplier No. 69292688
The Jackson Safety Internal Safety Lens is a genuine replacement part designed for use with Jackson TrueSight and TrueSight II auto-darkening welding lenses. It protects the auto-darkening filter (ADF) from sparks, scratches, and smoke, extending its lifespan and maintaining clear visibility during welding operations.
  • Works with TrueSight and TrueSight II digital ADF welding helmets
  • Extends the life of your ADF filter
  • Helps protect from sparks, scratches, and smoke
  • Maintains clear visibility for welding tasks
  • Genuine replacement part