Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Jackson Safety Replacement Sweatband, Black, for 117A Headgear, 2/Pk.

Catalog No. 5000687282
Encompass

These replacement sweatbands are designed for use with Jackson Welding Helmets.

  • Accessory type: replacement sweatband
  • For use with 117A headgear
  • Color: black
  • Comes in a pack of two

Catalog No. 50-006-87282 Supplier MSC Supplier No. 54983630
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These replacement sweatbands are designed for use with Jackson Welding Helmets.

  • Accessory type: replacement sweatband
  • For use with 117A headgear
  • Color: black
  • Comes in a pack of two

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.