Learn More
MSC Jackson Safety Safety Glasses, Scratch-Resistant, Polycarbonate, Amber, Full-Framed, 1 ea
Catalog No. 5000769852
Encompass
Jackson Safety SG Series safety glasses are designed for eye protection in various work environments. These full-framed glasses feature scratch-resistant polycarbonate amber lenses and a black frame with non-adjustable temples, meeting ANSI Z87.1+, CE, and CSA Z94.3 standards.
  • Scratch-resistant polycarbonate lens material
  • Amber lenses for enhanced contrast
  • Full-framed design for robust protection
  • Black frame with non-adjustable temples
  • UV protection
  • Soft temples and flexible nose piece for comfort
  • Includes dispenser box and neck cord
  • Universal size
  • Nylon frame material
  • Flexible temple type
Catalog No. 50-007-69852 Supplier MSC Supplier No. 10068575
Jackson Safety SG Series safety glasses are designed for eye protection in various work environments. These full-framed glasses feature scratch-resistant polycarbonate amber lenses and a black frame with non-adjustable temples, meeting ANSI Z87.1+, CE, and CSA Z94.3 standards.
  • Scratch-resistant polycarbonate lens material
  • Amber lenses for enhanced contrast
  • Full-framed design for robust protection
  • Black frame with non-adjustable temples
  • UV protection
  • Soft temples and flexible nose piece for comfort
  • Includes dispenser box and neck cord
  • Universal size
  • Nylon frame material
  • Flexible temple type