Learn More
MSC Jackson Safety Safety Plate Kit, Polycarbonate, Clear, Nitro, 1 Kit
Catalog No. 5000781351
Encompass
The Jackson Safety Nitro Safety Plate Lens Kit is designed for welding applications. It features durable polycarbonate lenses that offer ultimate protection, ensuring preparedness for welding jobs.
  • Kit includes 2 internal safety plates and 5 front safety plates
  • Made from durable polycarbonate material
  • Clear lens/plate color
  • Designed for Nitro welding models
  • Mount type: Internal clip-on
  • Overall height: 3.54 in
  • Overall width: 4.33 in
  • No lens shade
Catalog No. 50-007-81351 Supplier MSC Supplier No. 47574140
The Jackson Safety Nitro Safety Plate Lens Kit is designed for welding applications. It features durable polycarbonate lenses that offer ultimate protection, ensuring preparedness for welding jobs.
  • Kit includes 2 internal safety plates and 5 front safety plates
  • Made from durable polycarbonate material
  • Clear lens/plate color
  • Designed for Nitro welding models
  • Mount type: Internal clip-on
  • Overall height: 3.54 in
  • Overall width: 4.33 in
  • No lens shade