Learn More
MSC Jackson Safety Welding Cover Plates, Clear, Polycarbonate, Clip-On, 10 Sets
Catalog No. 5000782780
Encompass
Jackson Safety welding lenses and plates are designed to provide clear vision and protection in welding applications. These cover plates feature a clear polycarbonate construction with an internal clip-on mount for convenient use.
  • Clear polycarbonate material ensures optical clarity
  • Internal clip-on mount for secure and easy installation
  • Each set contains 7 pieces for comprehensive protection
  • Supplied in a pack of 10 sets for multiple applications
  • Features a window height of 2.36 in (5.99 cm)
Catalog No. 50-007-82780 Supplier MSC Supplier No. 68928241
Jackson Safety welding lenses and plates are designed to provide clear vision and protection in welding applications. These cover plates feature a clear polycarbonate construction with an internal clip-on mount for convenient use.
  • Clear polycarbonate material ensures optical clarity
  • Internal clip-on mount for secure and easy installation
  • Each set contains 7 pieces for comprehensive protection
  • Supplied in a pack of 10 sets for multiple applications
  • Features a window height of 2.36 in (5.99 cm)