Learn More
MSC Jackson Safety Welding Helmet, Thermoplastic, Auto-Darkening, Shade 9-13, 1 ea
Catalog No. 5000794190
Encompass
This welding helmet features a thermoplastic construction and an auto-darkening lens with a shade range of 9 to 13. It offers a fixed front window design for consistent protection during welding operations.
  • Thermoplastic is a lightweight polymer that is resistant to chemicals, heat, and scratches.
  • Auto-darkening lens with a shade range of 9 to 13.
  • Fixed front window style.
  • Powered by batteries with solar assist panel.
  • Green lens color.
  • Field of view measures 3.93 in width by 2.36 in height.
  • Features Arc Angel graphics.
  • Kit includes weld lens kit, shield and goggle combo, and poly clear insight weld lens cover.
Catalog No. 50-007-94190 Supplier MSC Supplier No. 37647757
This welding helmet features a thermoplastic construction and an auto-darkening lens with a shade range of 9 to 13. It offers a fixed front window design for consistent protection during welding operations.
  • Thermoplastic is a lightweight polymer that is resistant to chemicals, heat, and scratches.
  • Auto-darkening lens with a shade range of 9 to 13.
  • Fixed front window style.
  • Powered by batteries with solar assist panel.
  • Green lens color.
  • Field of view measures 3.93 in width by 2.36 in height.
  • Features Arc Angel graphics.
  • Kit includes weld lens kit, shield and goggle combo, and poly clear insight weld lens cover.