Learn More
MSC Jackson Safety Welding Helmet, Thermoplastic, Auto-Darkening, Shade 9-13, 1 ea
Catalog No. 5000792598
Encompass
This welding helmet features a lightweight thermoplastic construction and an auto-darkening lens with an adjustable shade range of 9 to 13. It is designed for various welding applications, offering both protection and enhanced visibility.
  • Auto-darkening lens with adjustable shade from 9 to 13
  • Lightweight thermoplastic material for durability
  • Resistant to chemicals, heat, and scratches
  • Fixed front window style
  • Powered by batteries with solar assist panel
  • Green lens color for clear vision
  • Field of view width: 3.93 in, height: 2.36 in
  • Arc Angel helmet graphics
Catalog No. 50-007-92598 Supplier MSC Supplier No. 37647740
This welding helmet features a lightweight thermoplastic construction and an auto-darkening lens with an adjustable shade range of 9 to 13. It is designed for various welding applications, offering both protection and enhanced visibility.
  • Auto-darkening lens with adjustable shade from 9 to 13
  • Lightweight thermoplastic material for durability
  • Resistant to chemicals, heat, and scratches
  • Fixed front window style
  • Powered by batteries with solar assist panel
  • Green lens color for clear vision
  • Field of view width: 3.93 in, height: 2.36 in
  • Arc Angel helmet graphics