Learn More
Med Vet International Jacobs Pin Chuck Only, J0214L, 10.5in, Each
Catalog No. 5000581655
Encompass_Preferred
Unique design on the standard instrument for introducing bone pins. An open slot cut into the handle permits visibilityof the pin resting in the chuck, saving time by allowing the surgeon to check pin depth without having to remove the chuck. The stainless steel Jacob’s accepts K-wires assmall as 0.035" and pins as large as 1/4". The handle and extension piece are lightweight, blue anodized aluminum alloy. Key and extension piece included.
Catalog No. 50-005-81655 Supplier Med Vet International Supplier No. JOR214L
Unique design on the standard instrument for introducing bone pins. An open slot cut into the handle permits visibilityof the pin resting in the chuck, saving time by allowing the surgeon to check pin depth without having to remove the chuck. The stainless steel Jacob’s accepts K-wires assmall as 0.035" and pins as large as 1/4". The handle and extension piece are lightweight, blue anodized aluminum alloy. Key and extension piece included.