Description
- Scissor-like ring handles
- Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
- Delicate hemostats have thin jaws with fine serrations.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Length (English) | 5 in. |
| Model | Straight, Extra-delicate |