Learn More
Stoelting™ Jacobson Mosquito Forceps
SDP
Catalog No. 10001327 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved, Extra-delicate
Straight, Extra-delicate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-327 Curved, Extra-delicate
10-001-326 Straight, Extra-delicate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-327 Supplier Stoelting™ Supplier No. 5211007
Add to Cart
Add to Cart
Stoelting™ Jacobson Mosquito Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
  • Delicate hemostats have thin jaws with fine serrations.
Specifications
For Use With (Application) Neuroscience
Length (English) 5 in.
Model Curved, Extra-delicate