Learn More
Med Vet International Jacquette 6s-7s Universal Sickle Scaler, Purple Handle, J0973EN, Each
Catalog No. 5000584000
Encompass_Preferred
Jacquette 6s-7s Universal Scaler: Thin, contra-angle scaler with offset bend for interproximal areas and anterior surfaces. Limited supply, no back orders, no returns.
Catalog No. 50-005-84000 Supplier Med Vet International Supplier No. JOR973EN
Jacquette 6s-7s Universal Scaler: Thin, contra-angle scaler with offset bend for interproximal areas and anterior surfaces. Limited supply, no back orders, no returns.