Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jaece Industries Identi-plug™ Plastic Foam Stoppers

Catalog No. 1412740C
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-40C White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-40C Supplier Jaece Industries Supplier No. L800B2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Autoclavable Autoclavable
Product Type Plug Stopper
Diameter (Metric) Inner 2.6 cm
Length (Metric) 4.44 cm
Color White
For Use With (Equipment) For 2.1 to 2.6cm test tubes and flasks openings
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.