Learn More
Fisherbrand™ Slide Holder
Catalog No. 1412750A
Catalog No. 14-127-50A Supplier Fisherbrand™ Supplier No. L500C
Add to Cart
Add to Cart
Provides an inexpensive way for drying excess fluids from flat surfaces. Fisherbrand™ Slide Holder is constructed of chemical-resistant HDPE.
Chemical-resistant high-density polyethylene holder
  • Convenient-to-use receptacle holds both 3 x 1 in. or 75 x 25 mm slides during evacuation and cleaning
  • Light, strong, chemical-resistant high-density polyethylene holder provides an inexpensive way for drying excess fluids from flat surfaces
  • 10° angle of slots facilitates clear viewing of desired slide
  • Beveled edges provide easy access to channels