Learn More
Medchemexpress LLC HY-P1846A 1mg Medchemexpress, Jagged-1 (188-204) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1846A1MG
Encompass_Preferred
Medchemexpress, HY-P1846A 1mg Jagged-1 (188-204) (TFA) CAS: Jagged-1 (188-204) TFA is a fragment of the Jagged-1 (JAG-1) protein. JAG-1 is a Notch ligand highly expressed in cultured and primary multiple myeloma (MM) cells. JAG-1 induces maturation of monocyte-derived human dendritic cells [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4249