Learn More
AdipoGen Jagged-2 hFc h rec
SDP
Catalog No. 502562968
Encompass_Preferred
Protein. Human Jagged-2 aa 1-1054 is fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02 % CHAPS. Purity >90% SDS-PAGE. Jagged-2 is a putative Notch ligand involved in the mediation of Notch signaling, that can induce stromal cells to secrete IL-6, VEGF and IGF-1. Notch activation can interact with NF-kappaB and C-myc to promote the proliferation and to inhibit the apoptosis of MM cells, showing in the relationship between the incidence of myeloma and drug resistance. Jagged-2 is also involved in limb development.
Catalog No. 50-256-2968 Supplier AdipoGen Supplier No. AG40A0155YC010
Protein. Human Jagged-2 aa 1-1054 is fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02 % CHAPS. Purity >90% SDS-PAGE. Jagged-2 is a putative Notch ligand involved in the mediation of Notch signaling, that can induce stromal cells to secrete IL-6, VEGF and IGF-1. Notch activation can interact with NF-kappaB and C-myc to promote the proliferation and to inhibit the apoptosis of MM cells, showing in the relationship between the incidence of myeloma and drug resistance. Jagged-2 is also involved in limb development.