Learn More
AdipoGen anti-Jagged-1 (human), mAb (J1G53-3) (100 µg)
SDP
Supplier:  AdipoGen AG20A0049C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human Jagged-1. Detects a band of ~180kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: J1G53-3. Applications: ELISA, FACS, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Jagged-1 is a ligand for multiple Notch receptors and involved in the mediation of Notch signaling. It is involved in cell-fate decisions during hematopoiesis. Is involved in early and late stages of mammalian cardiovascular development. Inhibits myoblast differentiation. Enhances fibroblast growth factor-induced angiogenesis (in vitro). Defects in Jagged-1 are the cause of Alagille syndrome type 1 (ALGS1) and tetralogy of Fallot (TOF).
Catalog No. 50-205-2849