Learn More
Medchemexpress LLC Jak/hdac-in-1 | 2284621-75-4 | 98.6% | 450.32 g/mol | C19H21Cl2N7O2 | 100MG
SDP
Catalog No. 5000239635
Encompass_Preferred
JAK/HDAC-IN-1 is a small-molecule dual inhibitor targeting JAK2 and histone deacetylase (HDAC) enzymes. It has reported low-nanomolar activity and shows antiproliferative and proapoptotic effects in hematological cell lines. The compound is supplied for research use only and is typically offered in small-mass quantities for in vitro studies.
Catalog No. 50-002-39635 Supplier Medchemexpress LLC Supplier No. HY126141100MG
JAK/HDAC-IN-1 is a small-molecule dual inhibitor targeting JAK2 and histone deacetylase (HDAC) enzymes. It has reported low-nanomolar activity and shows antiproliferative and proapoptotic effects in hematological cell lines. The compound is supplied for research use only and is typically offered in small-mass quantities for in vitro studies.