Learn More
Medchemexpress LLC Jak/HDAC-in-1 | 2284621-75-4 | 450.32 g·mol⁻¹ | C19H21Cl2N7O2 | 10MG
SDP
Catalog No. 5000239576
Encompass_Preferred
JAK/HDAC-IN-1 is a small-molecule dual inhibitor of Janus kinase 2 (JAK2) and histone deacetylases (HDAC) with low-nanomolar activity in biochemical assays. It exhibits antiproliferative and proapoptotic effects in several hematological cell lines and is supplied as a research reagent for in vitro studies.
  • Potent dual inhibitor of JAK2 and HDAC.
  • Low-nanomolar potency in biochemical assays.
  • Demonstrates antiproliferative and proapoptotic effects in hematological cell lines.
  • Available in small pack sizes suitable for in vitro research.
  • Molecular formula C19H21Cl2N7O2; molecular weight 450.32 g·mol⁻¹.
  • For research use only; not for human consumption or therapeutic use.
Catalog No. 50-002-39576 Supplier Medchemexpress LLC Supplier No. HY12614110MG
JAK/HDAC-IN-1 is a small-molecule dual inhibitor of Janus kinase 2 (JAK2) and histone deacetylases (HDAC) with low-nanomolar activity in biochemical assays. It exhibits antiproliferative and proapoptotic effects in several hematological cell lines and is supplied as a research reagent for in vitro studies.
  • Potent dual inhibitor of JAK2 and HDAC.
  • Low-nanomolar potency in biochemical assays.
  • Demonstrates antiproliferative and proapoptotic effects in hematological cell lines.
  • Available in small pack sizes suitable for in vitro research.
  • Molecular formula C19H21Cl2N7O2; molecular weight 450.32 g·mol⁻¹.
  • For research use only; not for human consumption or therapeutic use.