Learn More
Medchemexpress LLC Jak-in-5 (hydrochloride) | 2751323-21-2 | 99.5% | 511.03 | 100 MG
SDP
Catalog No. 5000346164
Encompass_Preferred
JAK-IN-5 hydrochloride is an inhibitor of JAK, extracted from patent US20170121327A1, compound example 283. It is for research use only.
  • Inhibits STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction.
  • Inhibits 88% of BALF eosinophils in male C57 mice model of Alternaria alternata-induced eosinophilic inflammation of the lung.
Catalog No. 50-003-46164 Supplier Medchemexpress LLC Supplier No. HY111471A100MG
JAK-IN-5 hydrochloride is an inhibitor of JAK, extracted from patent US20170121327A1, compound example 283. It is for research use only.
  • Inhibits STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction.
  • Inhibits 88% of BALF eosinophils in male C57 mice model of Alternaria alternata-induced eosinophilic inflammation of the lung.