Learn More
Medchemexpress LLC JAK-IN-5 hydrochloride | 2751323-21-2 | 99.5% | 511.03 | 25 MG
SDP
Catalog No. 5000346759
Encompass_Preferred
JAK-IN-5 hydrochloride is an inhibitor of JAK that was extracted from patent US20170121327A1, specifically as compound example 283. It has been shown to inhibit STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction. Additionally, in a male C57 mouse model of Alternaria alternata-induced eosinophilic inflammation of the lung, JAK-IN-5 hydrochloride inhibited 88% of BALF eosinophils.
  • Inhibitor of JAK.
  • Inhibits STAT6 phosphorylation in a mouse model.
  • Reduces BALF eosinophils in a mouse model of inflammation.
  • For research use only.
Catalog No. 50-003-46759 Supplier Medchemexpress LLC Supplier No. 25MGHY111471A
JAK-IN-5 hydrochloride is an inhibitor of JAK that was extracted from patent US20170121327A1, specifically as compound example 283. It has been shown to inhibit STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction. Additionally, in a male C57 mouse model of Alternaria alternata-induced eosinophilic inflammation of the lung, JAK-IN-5 hydrochloride inhibited 88% of BALF eosinophils.
  • Inhibitor of JAK.
  • Inhibits STAT6 phosphorylation in a mouse model.
  • Reduces BALF eosinophils in a mouse model of inflammation.
  • For research use only.