Learn More
Medchemexpress LLC JAK-IN-5 hydrochloride | 2751323-21-2 | 99.5% | 511.03 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY111471A10MG
Encompass_Preferred
JAK-IN-5 hydrochloride is an inhibitor of JAK, extracted from patent US20170121327A1. It is intended for research use only.
  • Inhibits JAK.
  • Inhibits STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction at 0.5 mg/mL (oral aspirate, 50 μL).
  • Inhibits 88% of BALF eosinophils in a male C57 mice model of Alternaria alternata-induced eosinophilic inflammation of the lung at 0.1-1.0 mg/mL (oral aspirate, 50 μL).
Catalog No. 50-003-46071