Learn More
Medchemexpress LLC JAK-IN-5 hydrochloride | 2751323-21-2 | 99.52% | 511.03 | 100 MG
SDP
Catalog No. 5000346024
Encompass_Preferred
JAK-IN-5 hydrochloride is an inhibitor of JAK extracted from patent US20170121327A1, compound example 283.
  • Inhibitor of JAK.
  • Inhibits STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction (oral aspirate, 0.5 mg/mL, 50 μL).
  • Inhibits 88% of BALF eosinophils in a male C57 mice model of Alternaria alternata-induced eosinophilic inflammation of the lung (oral aspirate, 0.1-1.0 mg/mL, 50 μL).
Catalog No. 50-003-46024 Supplier Medchemexpress LLC Supplier No. HY111471A50MG
JAK-IN-5 hydrochloride is an inhibitor of JAK extracted from patent US20170121327A1, compound example 283.
  • Inhibitor of JAK.
  • Inhibits STAT6 phosphorylation by 60% in IL-13-induced lung tissue in a mouse model of pSTAT6 induction (oral aspirate, 0.5 mg/mL, 50 μL).
  • Inhibits 88% of BALF eosinophils in a male C57 mice model of Alternaria alternata-induced eosinophilic inflammation of the lung (oral aspirate, 0.1-1.0 mg/mL, 50 μL).