Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JAK-IN-5 hydrochloride | 2751323-21-2 | 99.9% | 511.03 g/mol | C27H32ClFN6O | 5 MG
SDP

Catalog No. 5000263302
Encompass_Preferred

JAK-IN-5 hydrochloride is the hydrochloride salt of a Janus kinase (JAK) inhibitor disclosed in patent US20170121327A1 (compound example 283). Supplied for research use, it is characterized by high purity, defined solubility properties, and recommended storage conditions suitable for biochemical and cellular studies.

  • High purity: 99.86% (analytical grade).
  • Hydrochloride salt form for improved stability and handling.
  • Soluble in DMSO (5 mg/mL); sparingly soluble in water (0.5 mg/mL; sonication or warming may be required).
  • Storage: 4°C sealed; in solution: -80°C (6 months), -20°C (1 month).
  • Intended for research use as a JAK family kinase inhibitor.
  • Molecular weight 511.03 g/mol; molecular formula C27H32ClFN6O.

Catalog No. 50-002-63302 Supplier Medchemexpress LLC Supplier No. HY111471A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JAK-IN-5 hydrochloride is the hydrochloride salt of a Janus kinase (JAK) inhibitor disclosed in patent US20170121327A1 (compound example 283). Supplied for research use, it is characterized by high purity, defined solubility properties, and recommended storage conditions suitable for biochemical and cellular studies.

  • High purity: 99.86% (analytical grade).
  • Hydrochloride salt form for improved stability and handling.
  • Soluble in DMSO (5 mg/mL); sparingly soluble in water (0.5 mg/mL; sonication or warming may be required).
  • Storage: 4°C sealed; in solution: -80°C (6 months), -20°C (1 month).
  • Intended for research use as a JAK family kinase inhibitor.
  • Molecular weight 511.03 g/mol; molecular formula C27H32ClFN6O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.